Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Magnesium Green™, Pentapotassium Salt, cell impermeant

Catalog No. M3733
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M3733 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M3733 Supplier Invitrogen™ Supplier No. M3733
  • $476.00
    $390.32 / Each of 1

    Save $85.68

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Exhibits higher affinity for Mg2+ (Kd approximately 1.0mM) than mag-fura-2 (Kd approximately 1.9mM) or mag-indo-1 (Kd approximately 2.7mM)

 

  • Upon binding Mg2+, Magnesium Green exhibits an increase in fluorescence emission intensity without shift in wavelength
  • Absorption/emission maxima approximately 506/531nm

 

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Magnesium Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope, Microplate Reader
Product Type Pentapotassium Salt
Dye Type Magnesium Indicator
Product Line Magnesium Green
Quantity 1 mg
Shipping Condition Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.