Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Oregon Green™ 488 BAPTA-1, Hexapotassium Salt, cell impermeant

Catalog No. O6806
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O6806 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O6806 Supplier Invitrogen™ Supplier No. O6806
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

A visible light-excitable calcium indicator

Oregon Green 488 BAPTA-1, hexapotassium salt is a visible light—excitable calcium indicator. This water-soluble, salt form is useful for intracellular loading by microinjection, infusion from patch pipette or uptake induced by our Influx pinocytic cell-loading reagent (I-14402). The cell-permeant AM ester form of the indicator is also available (O-6807).

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Calcium Indicator
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer, Microplate Reader
Product Type Calcium Indicator
Dye Type Oregon Green
Excitation Wavelength Range 480 to 495 nm
Product Line Oregon Green
Quantity 500 μg
Shipping Condition Room Temperature
Emission Wavelength Range 510 to 525 nm
Excitation Wavelength 494 nm
Emission Wavelength 523 nm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.