Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OxyBURST™ Green H2HFF BSA - Special Packaging

Catalog No. O13291
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O13291 5 x 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O13291 Supplier Invitrogen™ Supplier No. O13291
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Sensitive fluorogenic reagent for detecting extracellular release of oxidative products in a spectrofluorometer/fluorescence microscope

Cell Analysis, Cell Cycle, Cell Structure, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Lysosomes, Vacuoles

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Product Type Supplement
Form Solid
Indicator Free Radical Sensor
Indicator Properties Fluorescent Dye-Based
Product Line OXYBURST
Quantity 5 x 1 mg
Reagent Type Fluorogenic
Shipping Condition Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.