Learn More
Invitrogen™ Multiplexed Proteomics™ Phosphoprotein Gel Stain Kits (with Pro-Q™ Diamond and SYPRO™ Ruby Gel Stains)
Description
Our new Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis. This reagent, when combined with our total protein stain, SYPRO Ruby Protein Gel Stain, becomes a powerful combination for quantitative proteome analysis.
Features include:
• Sensitive—Pro-Q Diamond Phosphoprotein Gel Stain offers detection level down to 1–16 ng of phosphoprotein per band
• Convenient—selectively stain phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein-specific antibodies and western blot analysis. SYPRO Ruby Gel Stain is subsequently used for total protein staining and simplifies the determination of phosphorylation level
• Linear signal—quantify with ease as Pro-Q Diamond Phosphoprotein Gel Stain and SYPRO Ruby Gel Stain exhibit excellent linearity over three orders of magnitude, ideal for quantitation
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | • Pro-Q Diamond Phosphoprotein Gel Stain, 200 mL • SYPRO Ruby Protein Gel Stain, 200 mL Store at room temperature and protect from light. |
| Description | Multiplexed Proteomics™ Phosphoprotein Gel Stain Kit #2 |
| Detection Location | In-Gel Detection |
| Detection Method | Fluorescence |
| Label or Dye | SYPRO Ruby, Pro-Q Diamond |
| Product Type | Phosphoprotein Gel Stain Kit |
| Product Line | PRO-Q™, SYPRO™ |
| Quantity | 1 Set, 200 mL per Dye |
| Shipping Condition | Room Temperature |
| Target Molecule | Proteins (Phosphoproteins) |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.