Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RediPlate™ 96 EnzChek™ Tyrosine Phosphatase Assay Kit

Catalog No. R22067
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 x 96-well plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R22067 1 x 96-well plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R22067 Supplier Invitrogen™ Supplier No. R22067
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready-to-use microplate assay

The RediPlate 96 EnzChek Tyrosine Phosphatase Assay Kit is a ready-to-use, fluorescence-based microplate assay for detecting tyrosine phosphatases (PTPase) and their corresponding modulators and inhibitors. Each RediPlate 96-well microplate has removable lanes, enabling researchers to perform only as many assays as needed for each experiment or to perform high-throughput analysis.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Enzyme and Metabolic Activity
For Use With (Equipment) Microplate Reader
Product Type Phosphatase Assay
Dye Type DiFMU
Excitation Wavelength Range 345 to 360 nm
Product Line EnzChek
Quantity 1 x 96-well plate
Shipping Condition Room Temperature
Substrate DiFMUP
Substrate Properties Chemical Substrate
Substrate Type Phosphatase Substrate
Target Enzyme Tyrosine Phosphatases
Emission Wavelength Range 450 to 465 nm
Excitation Wavelength 358 nm
Emission Wavelength 452 nm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.