Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rhod-2, Tripotassium Salt, cell impermeant

Catalog No. R14220
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R14220 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R14220 Supplier Invitrogen™ Supplier No. R14220
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cells may be physically loaded with cell-impermeant salt forms of these indicators using patch pipet, microinjection or pinocytotic cell-loading reagent.

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+. They have uses in many calcium signaling investigations, including measuring Ca2+ in cells and tissues that have high levels of autofluorescence and also for detecting Ca2+ release generated by photoreceptors and photoactivatable chelators. Cells may be physically loaded with the cell-impermeant salt forms of these indicators using patch pipette, microinjection, or our Influx™ pinocytotic cell-loading reagent (Cat. No. I14402). The fluorescence signal from these cells is generally measured using fluorescence microscopy.

Calcium Indicator (Cell-Impermeant Salts) Specifications:

  • Label (Ex/Em of Ca2+–bound form): Rhod-2 (552/581 nm)
  • Fluorescence intensity increase upon binding Ca2+: >100 fold
  • Kd for Ca2+ in the absence of Mg2+, in buffer: ∼570 nM
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength

Using TPEN to Control Heavy Metal Cations

In addition, BAPTA-based indicators such as these bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+. Perturbations to calcium measurements caused by presence of these ions can be controlled using the heavy metal-selective chelator (Cat. No. TPEN).

More Choices for Fluorescent Calcium Indicators

We offer a large selection of Molecular Probes™ calcium indicators for use in various experimental scenarios. For more information, review Fluorescent Ca2+ Indicators Excited with Visible Light—Section 19.3 in the Molecular Probes™ Handbook.

For UV-excitable Ca2+ indicators, protein-based Ca2+ indicators, conjugates of Ca2+ indicators, and for fluorescence-based indicators of other metal ions (i.e., Mg2+, Zn2+) review Indicators for Ca2+, Mg2+, Zn2+ and Other Metal Ions—Chapter 19 in the Molecular Probes™ Handbook.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Calcium Indicator
For Use With (Equipment) Fluorescence Microscope
Product Type Calcium Indicator
Dye Type Rhod-2
Quantity 1 mg
Shipping Condition Room Temperature
Excitation Wavelength 552 nm
Emission Wavelength 581 nm
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.