Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SNARF™-1 Carboxylic Acid, Acetate, Succinimidyl Ester - Special Packaging

Catalog No. S22801
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S22801 10 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S22801 Supplier Invitrogen™ Supplier No. S22801
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Permits simultaneous long-term tracing of mixed-cell populations using different fluorescent colors

To permit simultaneous long-term tracing of mixed-cell populations using different fluorescent colors, we have developed a probe whose applications should be similar to those of CFDA SE (commonly called CFSE). Cells labeled with our SNARF-1 carboxylic acid, acetate, succinimidyl ester have orange-red fluorescence that can be easily distinguished from that of cells loaded with green-fluorescent tracers such as CFDA SE.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C.
Detection Method Fluorescence
For Use With (Application) Cell Tracing
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer, Microplate Reader
Product Type Carboxylic Acid Acetate Succinimidyl Ester
Dye Type Fluorescent Dye-Based
Indicator pH Indicator
Product Line SNARF
Quantity 10 x 50 μg
Shipping Condition Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.