Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SphingoStrips™ Membranes

Catalog No. S23753
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S23753 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S23753 Supplier Invitrogen™ Supplier No. S23753
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

For protein identification

SpingoStrips™ membranes are designed for the identification of proteins possessing phosphoinositide or sphingolipid recognition domains and for analysis of their lipid-binding specificities.

Cell Analysis, Cell Structure, Enzyme and Protein Activity Assays, Lipase and Phopholipase Activity, Membranes (General) and Lipids, Plasma Membrane, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator 2°C to 8°C and protect from light.
Product Type Lipid Arrays
Format Membrane
Product Line SphingoStrips
Quantity 1 set
Shipping Condition Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.