Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TC-FlAsH™ II In-Cell Tetracysteine Tag Detection Kit (Green Fluorescence), for live-cell imaging

Catalog No. T34561
Encompass_Preferred
Click to view available options
Quantity:
1 Kit

Includes

Kit contains FlAsH-EDT2 labeling reagent and BAL wash buffer.

Contains an expression tag-based fluorescence labeling reagent for live-cell labeling

Mammalian cell lines expressing a protein fused to a tetracysteine tag (CCPGCC) can be labeled with the green-fluorescent FlAsH-EDT2 reagent contained in the kit. The tagged protein is fluorescent only when the labeling reagent is added. BAL wash buffer replaces the previously supplied Disperse Blue 3 and EDT-based wash buffer as an olfactorily more agreeable reagent that yields superior signal to noise.

Cell Analysis, Cell-Based Reporter Assays, Cellular Imaging, Enzyme and Protein Activity Assays, Labeling Chemistry, Lumio Detection, Proteins, Expression, Isolation and Analysis, Reporter Assays, Tetracysteine (Lumio/FlAsH/ReAsH) Assays

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Tetracysteine Tag Detection Kit
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Assay Tetracysteine Tag Detection
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Color Green
Label or Dye FlAsH
Shipping Condition Wet Ice
Format Kit
Product Line TC-FlAsH II
Quantity 1 Kit
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.