Learn More
Invitrogen™ Transferrin From Human Serum, Alexa Fluor™ 568 Conjugate
Description
Transferrin is a monomeric serum glycoprotein (approx. 80,000 daltons) that binds to a specific receptor on the surface of vertebrate cells and delivers up to two Fe3+ atoms via receptor-mediated endocytosis.
- Once iron-carrying transferrin proteins are inside endosomes, the acidic environment favors dissociation of iron from the transferrin-receptor complex
- Following release of iron, apotransferrin is recycled to plasma membrane, where it is released from its receptor to scavenge more iron
- Fluorescent transferrin conjugates can therefore be used with fluorescent LDL to distinguish lysosomally directed and recycling endosomal pathways
- Experiments are typically performed by adding fluorescently labeled transferrin to cultured cells and analyzing them by microscopy
- Label (Ex/Em): Alexa Fluor 594 (590/617)
- Amount: 15mg solid (contains 5mg of transferrin conjugate)
- Applications include: Image transferrin receptor dynamics using FRET; Observe receptor trafficking in live cells by confocal laser-scanning microscopy; Investigate events occurring during endosomal acidification; Measure transferrin receptor binding affinity in mammals and parasites
Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Chromatin Biology, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection, RNAi, Epigenetics and Non-Coding RNA Research
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Detection Method | Fluorescence |
| Product Type | Transferrin |
| Dye Type | Alexa Fluor Dyes |
| Form | Solid |
| pH | 7.2 |
| Product Line | Alexa Fluor |
| Quantity | 5 mg |
| Shipping Condition | Room Temperature |
| Excitation/Emission | 578/603 |
| Show More |
Frequently Asked Questions (FAQs)
Transferrin From Human Serum, Alexa Fluor 568 Conjugate is holotransferrin (saturated with iron). Lysine residues of transferrin are used for conjugation of the fluorophore. On the Certificate of Analysis, you can find information regarding the degree of labeling (number of fluorophores per one protein molecule).
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.