Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Valinomycin

Catalog No. V1644
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V1644 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V1644 Supplier Invitrogen™ Supplier No. V1644

Commonly used as an agent to induce apoptosis and can calibrate potentiometric responses in membrane experiments involving potential-sensitive dyes

  • Valinomycin is a natural, lipid-soluble molecule (Mr = approximately 1110) that binds potassium ions and facilitates their transfer across lipid bilayers (i.e., it is an ionophore)
  • A macrocyclic molecule formed from 12 alternating amino acids and esters, synthesized by specific Streptomyces
  • Exhibits a 10,000-fold selectivity for K+ over Na+
  • Valinomycin is a potent antibiotic
  • K+ selective ionophore
  • Neutrally charged
  • Soluble in ethanol, DMSO, etc., but insoluble in water

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Sodium and Potassium Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Product Type Valinomycin
Dye Type Ionophore
Quantity 25 mg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.