Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Zymosan A S. cerevisiae BioParticles™, unlabeled

Catalog No. Z2849
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z2849 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Z2849 Supplier Invitrogen™ Supplier No. Z2849
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nonfluorescent products are useful in phagocytosis assays, both as controls and for custom labeling with reactive dyes

  • Label (Ex/Em): Unlabeled
  • Particle: Zymosan (S. cerevisiae)
  • Provided as lyophilized powders
  • Approximately 3 x 108 S. aureus particles per mg solid and approximately 2 x 107 zymosan particles per mg solid
  • Conjugates can be reconstituted in the buffer of choice for use in phagocytosis assays

Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Flow Cytometry, Flow Cytometry Endocytosis, Exocytosis and Phagocytosis Assays, Flow Cytometry of Cellular Processes, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Ionic Homeostasis and Signaling, pH Detection

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type Bioparticle Conjugate
Dye Type Unconjugated
Form Solid
Product Line BioParticles
Quantity 100 mg
Shipping Condition Room Temperature
Species S. cerevisiae
Are the Invitrogen BioParticles products sterile?

While the bacteria have been attenuated with formaldehyde and alcohol desiccation, the BioParticles products are not considered sterile, and we do not recommend incubation of more than 4 hours. This applies to all of our dye-labeled (pHrodo, Alexa Fluor, etc.) and unlabeled BioParticles products.

What is the type of bond that attaches the dyes to the BioParticles probes?

We use amine-reactive dyes to covalently attach fluorescent dyes to all of our BioParticles probes such as the Escherichia coli (K-12 strain) BioParticles probes, Staphylococcus aureus (Wood strain without protein A) BioParticles, and the Zymosan A (S. cerevisiae) BioParticles probes.

What can the BioParticles product line be used for?

Fluorescent Bioparticles have been employed to study phagocytosis by fluorescence microscopy, quantitative spectrofluorometry, and flow cytometry. We offer E.Coli, S. aureus, and zymosan BioParticles conjugates covalently labeled with a variety of different fluorophores.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.