Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Molecular Sieve 5A Moisture Traps
Efficiently remove moisture and heavy hydrocarbons from compressed air, electrolytically produced hydrogen, house nitrogen, or other gases with high moisture or hydrocarbon content
Supplier: MilliporeSigma™ Supelco™ 20619
Description
- Moisture Traps (Drying Tubes) are suitable for maintaining anhydrous conditions within an apparatus
- Usually prepared by placing a small, loose plug of glass wool or cotton into the constriction at the end of the tube with a drying agent is poured on top
- Sieve 5A has the affinity for water and easily displaces other trapped gases
- Efficiently removes moisture and heavy hydrocarbons from compressed air, electrolytically produced hydrogen, house nitrogen, or other gases with high moisture or hydrocarbon content

Specifications
200 cc | |
Maintaining Anhydrous Conditions Within an Apparatus | |
26.5 in. | |
1 in. |
Moisture Trap | |
1/8 in. | |
67 cm | |
2.5 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction