Learn More
MilliporeSigma™ Supelco™ Molecular Sieve 5A Moisture Traps
Efficiently remove moisture and heavy hydrocarbons from compressed air, electrolytically produced hydrogen, house nitrogen, or other gases with high moisture or hydrocarbon content
$231.00 - $611.50
Specifications
Type Moisture Trap
For Use With (Application) Maintaining Anhydrous Conditions Within an Apparatus
View More Specs
Products 4
Catalog Number Mfr. No. Capacity (Metric) Size Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Size Price Quantity & Availability  
11-102-0825
View Documents
MilliporeSigma™ Supelco™
20618
200 cc 1/4 in.
Each for $231.00
Only null left
Add to Cart
11-102-0824
View Documents
MilliporeSigma™ Supelco™
20619
200 cc 1/8 in.
Each for $244.50
Only null left
Add to Cart
11-100-3821
View Documents
MilliporeSigma™ Supelco™
23992
750 cc 1/2 in.
Each for $611.50
Only null left
Add to Cart
11-101-6306
View Documents
MilliporeSigma™ Supelco™
23991
750 cc 1/4 in.
Each for $611.50
Only null left
Add to Cart
Description
Description
  • Moisture Traps (Drying Tubes) are suitable for maintaining anhydrous conditions within an apparatus
  • Usually prepared by placing a small, loose plug of glass wool or cotton into the constriction at the end of the tube with a drying agent is poured on top
  • Sieve 5A has the affinity for water and easily displaces other trapped gases
  • Efficiently removes moisture and heavy hydrocarbons from compressed air, electrolytically produced hydrogen, house nitrogen, or other gases with high moisture or hydrocarbon content
Specifications
Specifications
Moisture Trap
Maintaining Anhydrous Conditions Within an Apparatus
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.