Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Sieve, Type 3A, MilliporeSigma™

Catalog No. MMX1583D1 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MMX1583D1 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MMX1583D1 Supplier MilliporeSigma Supplier No. MX1583D1
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1313-59-3
Molecular Formula Na2O
Molecular Weight (g/mol) 61.979
InChI Key KKCBUQHMOMHUOY-UHFFFAOYSA-N
PubChem CID 73971
IUPAC Name disodium;oxygen(2-)
SMILES [O-2].[Na+].[Na+]

Specifications

Quantity 500 g
Packaging Glass Bottle
Physical Form Beads
Chemical Name or Material Molecular Sieve, Type 3A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.