Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Molecular sieves, 13X, 1-2mm (0.04-0.08 in.) dia.. pellets

Manufacturer:  Alfa Aesar™ 87954A7

Quantity 5kg

 View more versions of this product

Catalog No. AA87954A7


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

63231-69-6
MFCD00131613
Molecular sieves, 13X, 1 to 2mm (0.04 to 0.08 in.) dia. pellets

Specifications

1/16 in.
5kg
Pellet
Molecular sieves, 13X, 1 to 2mm (0.04 to 0.08 in.) dia. pellets
Hygroscopic
63231-69-6
MFCD00131613
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit