Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular sieves 3A, 8 to 12 mesh

Catalog No. AC197255000
Change view
Click to view available options
Quantity:
500 g
5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC197255000 500 g
AC197250050 5 kg
AC197250250 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC197255000 Supplier Thermo Scientific Chemicals Supplier No. 197255000
Only null left
Add to Cart

CAS: 308080-99-1 | AlNaO6Si2 | 202.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 308080-99-1
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.135
MDL Number MFCD00131613
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
IUPAC Name aluminum;sodium;dioxido(oxo)silane
SMILES [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]

Specifications

Color Beige to Brown
Quantity 500 g
Bulk Density 600g/L min.
Residual Water 1.5% max.
Packaging Glass bottle
Water Adsorption 20% min. (60% rel. humid.)
Physical Form Beads
Solubility Information Solubility in water: insoluble.
Chemical Name or Material Molecular sieves 3A, 8 to 12 mesh
Hazard Category HYGROSCOPIC

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.