Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular sieves, 5A, 3-4mm (0.12-0.16in) dia. pellets

Catalog No. p-7050041
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33551A1 1 kg
AA33551A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA33551A1 Supplier Thermo Scientific Chemicals Supplier No. 033551A1
Only null left
Add to Cart

CAS: 69912-79-4 | AlNaO6Si2 | 202.14 g/mol

Molecular sieves, 5A, 3-4mm (0.12-0.16in) dia. Pellets is used as an sorbent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69912-79-4
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.14
MDL Number MFCD00147627
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
IUPAC Name aluminum;sodium;dioxido(oxo)silane
SMILES [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O

Specifications

Color Brown
Quantity 1 kg
Sensitivity Hygroscopic
Physical Form Pellet
Solubility Information Insoluble in water.
Chemical Name or Material Molecular sieves 5A, 3 to 4mm (0.12 to 0.16 in.) dia. pellets
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.