Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Sieves, Grade 514 (Type 4A; 8-12 mesh), Fisher BioReagents

Catalog No. BP2631500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP2631500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP2631500
Used for dehydration of liquids.
TRUSTED_SUSTAINABILITY

Specifications

Color Beige
Chemical Name or Material Molecular Sieves, Grade 514
Density 2.1 g/cm3
Quantity 500 g
Mesh Size 8 to 12 Mesh
Identification Pass Test
Name Note Type 4A; 8-12 Mesh
Packaging Glass Bottle
Physical Form Solid
CAS 1318-02-1, 14808-60-7
Molecular Formula AlNaO6Si2
Synonym Aluminum Sodium Silicate
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
Molecular Weight (g/mol) 202.14
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.