Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Sieves, (Type 3A), Fisher Chemical™

Catalog No. M5645LB Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
5 lb.
Packaging:
Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M564-5LB 5 lb. Poly Bottle
M564-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M564-5LB

Specifications

Color Beige
Chemical Name or Material Molecular Sieves, Type 3A
pH 10.3 to 10.5
Quantity 5 lb.
Mesh Size 8 to 12 Mesh
Packaging Poly Bottle
Identification Pass test
Name Note Type 3A; 8-12 Mesh Beads
Physical Form Solid
CAS 12736-96-8, 1332-58-7, 14808-60-7
MDL Number MFCD00131613
Synonym Sodium Aluminosilicate, Aluminum Sodium Silicate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.