Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OPP (O-propargyl-puromycin)

Catalog No. C10459
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10459 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C10459 Supplier Invitrogen™ Supplier No. C10459
  • $1,914.00
    $1,569.48 / Each of 1

    Save $344.52

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Alkyne analog of puromycin

OPP (O-propargyl-puromycin) is an alkyne analog of puromycin that is efficiently incorporated into newly translated proteins in complete methionine-containing media. When used with Click-iT® Plus OPP Alexa Fluor® Protein Synthesis assays or Alexa Fluor® Picolyl Azide toolkits, OPP provides a fast, sensitive, and non-radioactive method for the detection of protein synthesis using fluorescence microscopy, high-content imaging, or flow cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and desiccate.
For Use With (Application) Fluorescence Microscopy, High-content Imaging, Flow Cytometry
Product Type OPP
Product Line OPP (O-propargyl-puromycin)
Quantity 5 mg
Reagent Type Nascent Protein Labeling Reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.