Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Molnupiravir
GREENER_CHOICESDP

Catalog No. 758650 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-865-0 50 mg
75-861-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 75-865-0 Supplier R&D Systems™ Supplier No. 7586/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Prodrug of antiviral nucleoside analog; inhibits SARS-CoV, SARS-CoV-2 and MERS-CoV infection

Molnupiravir is a broad-spectrum antiviral prodrug of the nucleoside analog β-D-N4-hydroxycytidine (NHC). Molnupiravir competes predominantly with CTP for incorporation; it induces lethal viral mutagenesis by increasing G to A and C to U transition mutations without inhibition of RNA synthesis in replicating coronaviruses, which prevents the development of viral resistance. In primary airway epithelial cell cultures, Molnupiravir dose-dependently inhibits SARS-CoV-2 infectious virus production; it also inhibits MERS-CoV and SARS-CoV with EC50 values in the submicromolar range. In mice infected with SARS-CoV or MERS-CoV, Molnupiravir improves pulmonary function, and reduces virus titer and body weight loss. Molnupiravir also displays in vivo efficacy against seasonal and pandemic influenza A shed virus. It is orally bioavailable.

Specifications

Chemical Name or Material β-D-N4-hydroxycytidine-5 '-isopropyl ester
CAS 2492423-29-5
Quantity 50 mg
Product Line Tocris
Target Antiviral Compounds
Molecular Formula C13H19N3O7
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.