Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Molybdenum

Catalog No. 503093918 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
10 Ea.
5 Ea.

Catalog No. 50-309-3918

Supplier: Sigma Aldrich GF447776161EA

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Molybdenum

Specifications

CAS 7439-98-7
Melting Point 2617°C
Density 10.3 g/mL (at 25°C)
Boiling Point 4612°C
Molecular Formula Mo
Linear Formula Mo
Quantity 1 Ea.
MDL Number MFCD00003465
Synonym Molybdenum; MO000310
Molecular Weight (g/mol) 95.94
Percent Purity 99.9%
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.