Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenum AA Standard, 1000 ppm Mo in 3% HCl/tr HF, Ricca Chemical
SDP

Catalog No. amo1kh100
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle

Chemical Identifiers

CAS 1313-27-5, 7647-01-0, 1313-27-5, 7664-39-3, 7732-18-5
Molecular Formula MoO3
Molecular Weight (g/mol) 143.95
MDL Number MFCD00003469
InChI Key JKQOBWVOAYFWKG-UHFFFAOYSA-N
PubChem CID 14802
ChEBI CHEBI:30627
IUPAC Name trioxomolybdenum
SMILES O=[Mo](=O)=O

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm
CAS Min % 94.0, 3.9, 0.14, 0.01
CAS Max % 98.0, 4.06, 0.15, 0.01
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Solvent or Matrix 3% HCl/tr HF
Quantity 100 mL
Chemical Name or Material Molybdenum AA Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.