Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Molybdenum boride, 99.5% (metals basis)

Catalog No. AA4368014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43680-14 25 g
AA43680-22 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43680-14 Supplier Thermo Scientific™ Supplier No. 4368014
Add to Cart

CAS: 12006-99-4 | BMo2 | 202.71 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12006-99-4
Molecular Formula BMo2
Molecular Weight (g/mol) 202.71
MDL Number MFCD01310400
InChI Key QROVMOADHGTXCA-UHFFFAOYSA-N
IUPAC Name dimolybdenumynylidyneborane
SMILES B#[Mo]#[Mo]

Specifications

Physical Form Solid
Quantity 25 g
Formula Weight 202.69
ShelfLife 5 Years
EINECSNumber 234-502-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.