Learn More
Molybdenum boride, 99% (metals basis), Thermo Scientific Chemicals
Description
Molybdenum borides have been used as hard coating materials for high temperature structural applications, and also in the form of MoB/CoCr cermet coatings. It is also used as a brazing material especially in electric components. Molybdenum boride has potential industrial applications such as abrasive, corrosion resistant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 100 g |
Solubility Information | it is soluble in water,paraffin oil,ether,benzene and other organic solvents. |
Formula Weight | 106.75 |
Percent Purity | 99% |
Assay | (metals basis) |
Chemical Name or Material | Molybdenum boride |
Safety and Handling
EINECSNumber | 234-492-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.