Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenum boride, 99% (metals basis)

Catalog No. p-7049041
Click to view available options
Quantity:
25 g
100 g

CAS: 12006-98-3 | BMo | 106.76 g/mol

Molybdenum borides have been used as hard coating materials for high temperature structural applications, and also in the form of MoB/CoCr cermet coatings. It is also used as a brazing material especially in electric components. Molybdenum boride has potential industrial applications such as abrasive, corrosion resistant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12006-98-3
Molecular Formula BMo
Molecular Weight (g/mol) 106.76
MDL Number MFCD00014219
InChI Key LGLOITKZTDVGOE-UHFFFAOYSA-N
Synonym molybdenum boride, molybdenum boride mob, molybdenumylidyneborane, molybdenum monoboride, molybdenum boride mono , mob, molybdenum boride, powder,-325 mesh trace metals basis
PubChem CID 82790
IUPAC Name boranylidynemolybdenum
SMILES B#[Mo]

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information it is soluble in water,paraffin oil,ether,benzene and other organic solvents.
Formula Weight 106.75
Percent Purity 99%
Assay (metals basis)
Chemical Name or Material Molybdenum boride
EINECSNumber 234-492-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.