Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Molybdenum hexacarbonyl, 98%
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 150.0°C |
| Color | White through Yellow |
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Physical Form | Crystalline Powder |
| Quantity | 10 g |
| Assay Percent Range | 98% |
| Linear Formula | Mo(CO)6 |
| Fieser | 02,287; 03,206; 04,346; 09,317; 10,274; 13,194; 01,140; 02,448; 03,325; 04,559; 05,736; 06,652; 07,68; 08,109; 09,113; 15,9 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in benzene, slightly soluble in ether and most oter common, organic solvents |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. |
| Precautionary Statement | GHS P Statement Rinse mouth. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortabl |
| EINECSNumber | 237-713-3 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction