Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenum ICP Standard, 50 ppm Mo, Ricca Chemical
SDP

Catalog No. PMO50N120 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PMO50N-120 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PMO50N-120 Supplier Ricca Chemical Company Supplier No. PMO50N120
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7439-98-7 , 7697-37-2 , 7664-39-3 , 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Packaging Natural Poly Bottle
CAS Min % 94.0, 4.24, 0.0042, 0.0042
CAS Max % 98.0, 4.41, 0.0044, 0.0044
Odor Odorless
Color Colorless
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
Quantity 120 mL
Grade Laboratory
Concentration or Composition (by Analyte or Components) 1mL = 0.05mg Mo (50ppm Mo)
Chemical Name or Material Molybdenum ICP Standard
Show More Show Less
ShelfLife 6 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.