Alfa Aesar™ Molybdenum(II) acetate dimer

Manufacturer: Alfa Aesar™ 1823903

Quantity 1g

 View more versions of this product

Catalog No. AA1823903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14221-06-8
    Formula Weight 428.06
    MDL Number MFCD00013395
    Chemical Name or Material Molybdenum(II) acetate dimer

    Specifications

    Formula Weight 428.06
    Quantity 1g
    Sensitivity Air Sensitive
    Physical Form Crystalline
    MDL Number MFCD00013395
    CAS 14221-06-8
    Chemical Name or Material Molybdenum(II) acetate dimer

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit