Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenum(IV) sulfide, 98%

Catalog No. AA4182722
Click to view available options
Quantity:
25 g
100 g
500 g

Catalog No. AA4182722

Supplier: Thermo Scientific Chemicals 04182722

Only null left
Add to Cart

CAS: 1317-33-5 | MoS2 | 160.07 g/mol

Molybdenum(IV) sulfide is used as a catalyst in the electrolysis of water. It is employed as a cocatalyst for desulfurization in petrochemistry especially hydrodesulfurization. It is also used as a component of photoelectrochemical and microelectronics applications. Further, it is useful as a filament material in many electrical applications. It finds application as a lubricant in aircraft engines. Polymers consisting of molybdenum(IV) sulfide such as nylon, teflon and vespel posses improved strength and reduced friction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-33-5
Molecular Formula MoS2
Molecular Weight (g/mol) 160.07
MDL Number MFCD00003470
InChI Key CWQXQMHSOZUFJS-UHFFFAOYSA-N
Synonym molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z
PubChem CID 14823
ChEBI CHEBI:30704
SMILES S=[Mo]=S

Specifications

Melting Point 1185°C
Color Black
Physical Form Powder
Quantity 100 g
Assay Percent Range 99% (metals basis)
Solubility Information Soluble in hot sulfuric acid,and aquaregia. Insoluble in water,concentrated sulfuric acid and dilute acid.
IUPAC Name bis(sulfanylidene)molybdenum
Formula Weight 160.08
Odor Odorless
Appearance Black powder
Chemical Name or Material Molybdenum(IV) sulfide
Show More Show Less
Hazard Category H332
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
Precautionary Statement P261-P271-P304+P340-P312
EINECSNumber 215-263-9
RTECSNumber QA4697000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.