Molybdenum Rod, 12.7mm (0.5 in.) dia., 99.95% (Metals basis), Alfa Aesar™

Approx. 3.8cm/50g

Manufacturer: Alfa Aesar 1003418

Quantity 50g

 View more versions of this product

Catalog No. AA1003418

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-98-7
    Physical Form Rod, 12.7mm (0.5 in.) dia.
    Assay Percent Range 99.95% (Metals basis)
    MDL Number MFCD00003465
    Chemical Name or Material Molybdenum

    Specifications

    Quantity 50g
    Color Dark gray
    Physical Form Rod, 12.7mm (0.5 in.) dia.
    Assay Percent Range 99.95% (Metals basis)
    MDL Number MFCD00003465
    CAS 7439-98-7
    Odor Odorless
    Appearance Dark gray rod
    Chemical Name or Material Molybdenum

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit