Learn More
Molybdenum(V) isopropoxide, 99+% (metals basis)
Description
Molybdenum(V) isopropoxide, it is used as a Precursor used in sol-gel process. It is used in nanomaterials and thin films. It is used as a solution based product for printed electronics. It acts a catalyst for selective isobutane oxidation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Black to Yellow |
| Linear Formula | Mo[OCH(CH3)2]5 |
| Quantity | 2 g |
| UN Number | UN3181 |
| Sensitivity | Moisture sensitive |
| Solubility Information | It is soluble in hydrocarbons,reacts with alcohols,ketones and esters. It reacts exothermally with water. |
| Formula Weight | 391.04 |
| Percent Purity | ≥99% |
| Physical Form | Powder |
| Assay | (metals basis) |
| Show More |
Safety and Handling
| Hazard Category | H228-H315-H319-H335 |
| Hazard Statement | GHS H Statement H228-H315-H319-H335 Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.