Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molybdenum(VI) fluoride

Catalog No. AA2271318
Click to view available options
Quantity:
50 g
Packaging:
CGA 330 valve cylinder

CAS: 7783-77-9 | F6Mo | 209.94 g/mol

Molybdenum(VI) fluoride plays an important role in the semiconductor industry and the nuclear industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packed in one-way cylinder w/CGA 330 valve
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-77-9
Molecular Formula F6Mo
Molecular Weight (g/mol) 209.94
MDL Number MFCD00014220
InChI Key RLCOZMCCEKDUPY-UHFFFAOYSA-H
Synonym molybdenum hexafluoride, molybdenum fluoride mof6 , oc-6-11, molybdenum fluoride mof6, unii-zlt5j123w3, mof6, molybdenum vi fluoride, molybdenum 6+ fluoride, acmc-1bii8
PubChem CID 82219
ChEBI CHEBI:30708
IUPAC Name hexafluoromolybdenum
SMILES F[Mo](F)(F)(F)(F)F

Specifications

Boiling Point 37°C
Melting Point 17°C
Color Colorless
Physical Form Liquid
Quantity 50 g
Linear Formula MoF6
UN Number UN3289
Merck Index 14,6237
Solubility Information Miscible with saturated hydrocarbons,chlorinated and fluorinated hydrocarbons.
Formula Weight 209.93
Packaging CGA 330 valve cylinder
Sensitivity Moisture sensitive
Density 2.3 g/mL
Chemical Name or Material Molybdenum(VI) fluoride
Show More Show Less
Hazard Category H314-H330-H335
Hazard Statement GHS H Statement
H330-H314-H318
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUID, CORROSIVE, INORGANIC, N.O.S.
EINECSNumber 232-026-5
RTECSNumber QA4681450
TSCA Yes
storageNote1
  • Packed in one-way cylinder w/CGA 330 valve
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.