Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Molybdenum(VI) sulfide, 97%

Catalog No. AA4735914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA47359-14 25 g
AA47359-22 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA47359-14 Supplier Thermo Scientific™ Supplier No. 4735914
Add to Cart

CAS: 12033-29-3 | MoS{3}

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12033-29-3
Molecular Formula MoS{3}
MDL Number MFCD00014226

Specifications

Physical Form Solid
Quantity 25 g
Formula Weight 192.13
Percent Purity 97%
ShelfLife 5 Years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.