Learn More
Molybdic acid, MoO3 85% min
Description
Molybdic acid is used as a precursor in the production of ammonium molybdate tetrahydrate and molybdenum(IV) sulfide. The mixture of molybdic acid in concentrated sulfuric acid is called as Froehde reagent, which finds application in analytical chemistry for the detection of alkaloids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Molybdic acid is used as a precursor in the production of ammonium molybdate tetrahydrate and molybdenum(IV) sulfide. The mixture of molybdic acid in concentrated sulfuric acid is called as Froehde reagent, which finds application in analytical chemistry for the detection of alkaloids.
Solubility
Soluble in dilute ammonium hydroxide, alkali and ammonia. Slightly soluble in water.
Notes
Incompatible with strong bases and strong oxidizing agents.

Specifications
Specifications
Quantity | 2500 g |
Solubility Information | Soluble in dilute ammonium hydroxide,alkali and ammonia. Slightly soluble in water. |
Concentration or Composition (by Analyte or Components) | MoO3 85% min |
Density | 3.1 |
Chemical Name or Material | Molybdic acid |
Safety and Handling
Hazard Category | H319-H335-H373 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P260-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P314-P501c |
EINECSNumber | 231-970-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.