Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ MOMBA
GREENER_CHOICESDP

Catalog No. 7926250 Shop All Novus Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-262-50 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 79-262-50 Supplier R&D Systems™ Supplier No. 7926/250

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective orthostatic agonist of hFFA2-DREADDs

MOMBA is a selective orthosteric agonist for human free fatty acid 2 (hFFA2) engineered receptors, designer receptors activated by designer drugs (DREADD), it shows no activity to wild-type hFFA2, hFFA3, and mouse FFA2. MOMBA inhibit forskolin-stimulated cAMP levels in cells expressing the hFFA2-DREADD receptor with improved potency and equivalent efficacy to Sorbic acid (Cat. No. 7119). In hFFA2-DREADD transgenic mice, MOMBA treatment significantly reduces gut transit and promotes concentration-dependent release of GLP-1 from colonic crypts. Orally bioavailable.

Specifications

Chemical Name or Material 4-Methoxy-3-methylbenzoic acid
CAS 6880-04-2
Quantity 250 mg
Product Line Tocris
Target DREADD Activators
Molecular Formula C9H10O3
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.