Learn More
Moment of Inertia
SDPSCI_ED
Catalog No. S05955
Click to view available options
:
Moment of Inertia set
Includes
Two wands of equal length and mass, two rings of equal diameter and mass, movable masses on one wand and one ring, instructions
Explore the concept of moment of inertia with wands and rings of equal mass but different distribution of mass.
  • One set has an even distributions of mass, the other a movable mass
  • See which wand twirls more easily and which ring moves more quickly down an inclined plane?
Specifications
Product Type Moment of Intertia Set
Includes Two wands of equal length and mass, two rings of equal dia. and mass, movable masses on one wand and one ring, instruction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.