Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MON1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562834
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562834 Supplier Invitrogen™ Supplier No. PA562834
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CLPKFNAAGF FHAHISYLEP DTDLCLLLVS TDREDFFAVS DCRRRFQERL RKRGAHLALR EALRTPYYSV AQVGIPDLRH FLYKSKSSGL FTS Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.

MON1A is a cytosolic protein that peripherally associates with the ER. MON1A plays an important role in membrane trafficking through the secretory apparatus. MON1A affects different steps in the secretory pathway including endoplasmic reticulum-to-Golgi traffic. MON1A encodes several isoforms with MW about 62, 55, 45 and 34 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MON1A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MON1A
Gene Accession No. Q86VX9
Gene Alias 2810468K17Rik; FLJ97088; MGC13272; MON1 homolog A; MON1 homolog A (yeast); MON1 homolog A, secretory trafficking associated; MON1 secretory trafficking family member A; Mon1a; RGD1308823; SAND1; Vacuolar fusion protein MON1 homolog A
Gene Symbols MON1A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MON1A. Recombinant protein control fragment (Product #RP-96426).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84315
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.