Learn More
Invitrogen™ MON1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109969
Description
Immunogen sequence: MHPGGGPSRA ERLELGLGRE RPAKAIFLHR RPGEGGGRER CLRCGHVCVR RGPGPREAVP SGRPRPDTLT PPWVRQRAVT GTFCASWTPL RNRRAQRMAT DMQRKRSSEC LDGTLTPSDG QSMERAESPT PGMAQGMEPG AGQEGAMFVH ARSYEDLTES EDGAASGDSH KEGTRGPPPL PTDMRQISQD FSELSTQLTG VARDLQEEML PGSSEDWLEP PGAVGRPATE PPREGTTEGD EEDATEAWRL.
MON1A is a cytosolic protein that peripherally associates with the ER. MON1A plays an important role in membrane trafficking through the secretory apparatus. MON1A affects different steps in the secretory pathway including endoplasmic reticulum-to-Golgi traffic. MON1A encodes several isoforms with MW about 62, 55, 45 and 34 kDa.
Specifications
| MON1A | |
| Polyclonal | |
| Unconjugated | |
| MON1A | |
| 2810468K17Rik; FLJ97088; MGC13272; MON1 homolog A; MON1 homolog A (yeast); MON1 homolog A, secretory trafficking associated; MON1 secretory trafficking family member A; Mon1a; RGD1308823; SAND1; Vacuolar fusion protein MON1 homolog A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84315 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.73 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86VX9 | |
| MON1A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human MON1A (Q86VX9). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.