Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MON1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109970
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109970 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109970 Supplier Invitrogen™ Supplier No. PA5109970
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WVTSKFELYT CLSPLVTKAG AILVVTKLLR WVKKEEDRLF IRYPPKYSTP PATSTDQAAH NGLFTGL.

MON1B (MON1 Homolog B, Secretory Trafficking Associated) is a Protein Coding gene. Among its related pathways areVesicle-mediated transportandRAB GEFs exchange GTP for GDP on RABs. An important paralog of this gene isMON1A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MON1B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MON1B
Gene Accession No. Q7L1V2
Gene Alias 5031407H10; 5033413H12Rik; HSRG1; HSV-1 stimulation-related gene 1 protein; HSV-I stimulating-related protein; Kiaa0872; MON1 homolog B; MON1 homolog b (yeast); MON1 homolog B, secretory trafficking associated; MON1 secretory trafficking family member B; MON1B; SAND2; SRG1; Vacuolar fusion protein MON1 homolog B
Gene Symbols MON1B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 481-547 of human MON1B (NP_055755.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22879, 307868
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.