Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MON1B Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571071
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1071 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-1071 Supplier Bethyl Laboratories, Inc Supplier No. A305713AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

MON1B (MON1 Homolog B, Secretory Trafficking Associated) is a Protein Coding gene. Among its related pathways areVesicle-mediated transportandRAB GEFs exchange GTP for GDP on RABs. An important paralog of this gene isMON1A.

Specifications

Antigen MON1B
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene MON1B
Gene Accession No. Q7L1V2
Gene Alias HSRG1, HSV-1 stimulation-related gene 1 protein, HSV-I stimulating-related protein, KIAA0872, MON1 homolog B, MON1 homolog B (yeast), MON1 secretory trafficking family member B, SAND2, SRG1
Gene Symbols MON1B
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22879
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.