Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MON1B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572016
Description
MON1B belongs to the MON1/SAND family.
Specifications
MON1B | |
Polyclonal | |
Unconjugated | |
MON1B | |
Vacuolar fusion protein MON1 homolog B; HSV-1 stimulation-related gene 1 protein; HSV-I stimulating-related protein; MON1B; HSRG1; KIAA0872; SAND2 | |
Rabbit | |
Protein A | |
RUO | |
22879, 270096 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.49 mg/mL | |
PBS with 0.09% sodium azide | |
Q7L1V2, Q8BMQ8 | |
MON1B | |
KLH conjugated synthetic peptide between 396-425 amino acids from the C-terminal region of human MON1B | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction