Learn More
Novus Biologicals™ Monkey Circulating immune complex (CIC) ELISA Kit (Colorimetric)
Shop All R&D Systems Products

Specifications
Specifications
| Assay Range | 0.312 - 20 ug/mL |
| Assay Sensitivity | 0.078 ug/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Plasma, Serum |
| Specificity | This assay has high sensitivity and excellent specificity for detection of monkey Circulating immune complex (CIC). No significant cross-reactivity or interference between monkey Circulating immune complex (CIC) and analogues was observed. |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Storage Requirements | Store at 4°C. |
| Target | Monkey |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.