Learn More
Specifications
Specifications
| Assay Range | 0.16 to 10 ng/mL |
| Assay Sensitivity | 0.09 ng/mL |
| Immunoassay Kit Format | Colorimetric Detection Kit |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Serum, Plasma, Cell Supernatant |
| Specificity | This kit recognizes Monkey FABP2/I-FABP in samples. No Significant cross-reactivity or interference between Monkey FABP2/I-FABP and analogues was observed. |
| Target Species | Monkey |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 2169 |
| Gene Symbol | FABP2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
