Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Monkey Galectin-1 ELISA Kit

Catalog No. p-7219856
Encompass_Preferred
Click to view available options
Quantity:
5 x 96 Tests
96 Tests

ELISA

Monkey Galectin-1 quantitates monkey Galectin-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant monkey Galectin-1.

Galectin-1 belongs to a large family of carbohydrate-binding proteins called lectins. Galectin-1can be either monomeric or homodimeric and is found in a wide variety of cells and tissue types. Galectin-1 can control cell growth, proliferation, induce apoptosis of activated T cells while it can also modulate cytokine secretion or inhibit pro-inflammatory cytokine production. Galectin-1 plays an important role in acute and chronic inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 14 kDa laminin-binding protein; 14 kDa lectin; AA410090; beta-galactoside binding protein; Beta-galactoside-binding lectin L-14-I; beta-galactoside-binding protein 14 kDa; DKFZp686E23103; GAL1; gal-1; Galaptin; Galbp; galectin 1; Galectin1; galectin-1; galectin-1-like protein; Gbp; HBL; H-Galectin-1; HLBP14; HPL; L14; L-14.5; Lactose-binding lectin 1; Lect14; Lectin galactoside-binding soluble 1; lectin, galactose binding, soluble 1; lectin, galactoside binding soluble 1; lectin, galactoside-binding, soluble, 1; Lgals1; pm12; Putative MAPK-activating protein PM12; RL 14.5; S-Lac lectin 1
Assay Range 0.2 to 50 ng/mL
Assay Sensitivity 0.2 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Galectin 1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.