Learn More
Invitrogen™ Monkey IL-10 ELISA Kit

Description
Includes
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey IL-10
Biotin-Conjugate anti-monkey IL-10 monoclonal antibody
Streptavidin-HRP
Monkey IL-10 Standard lyophilized, 500 U/ml upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
The Monkey Interleukin 10 (Mk IL-10) ELISA quantitates Mk IL-10 in monkey (baboon, cynomolgus) serum or cell culture medium. The assay will exclusively recognize both natural and recombinant monkey IL-10. Principle of the method The Monkey IL-10 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Specifications
Specifications
Accession Number | P79338 |
Assay Range | 3.9 to 250 U/mL |
Assay Sensitivity | 1.3 U/mL |
Conjugate | HRP |
Product Type | ELISA |
Sample Type | Serum, Supernatant |
Target Species | Monkey |
For Use With (Application) | ELISA |
For Use With (Equipment) | Colorimetric Microplate Reader |
Gene ID (Entrez) | 101024270, 102133450 |
Show More |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.