Learn More
Description
The Monkey IL-4 DuoSet ELISA kit provides the core components for developing sandwich ELISAs to detect and quantify monkey Interleukin 4 (IL-4) in biological samples.
- Assay Type: Solid Phase Sandwich ELISA
- Reactivity: Primate
- Assay Range: 3.12 to 200 pg/mL
- Sample Type: Cell culture supernates, serum, and plasma
- Assay Format: 96-well strip plate
- Detection Method: Colorimetric ELISA at 450 nm (TMB)
- Sample Volume: 100 μL
- Conjugate: Biotin
- Label: HRP
- Research Area: Cytokines and Receptors in Allergy and Asthma
Specifications
Specifications
| Product Type | Solid Phase Sandwich ELISA |
| Quantity | 1 Kit (for 15 Plates) |
| Species | Primate |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3565 |
| Product Line | R&D Systems |
| Synonym | B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Research Area | Immunology |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
