Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Monkey Protein S/PROS1 ELISA Kit (Colorimetric)
SDP

Catalog No. NB398007 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398007 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398007 Supplier Novus Biologicals™ Supplier No. NBP308173
Add to Cart
Add to Cart

Kit (Colorimetric)

Specifications

Assay Range 1.56 to 100 ng/mL
Assay Sensitivity 0.94 ng/mL
Immunoassay Kit Format Colorimetric Detection Kit
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma
Target Species Monkey
For Use With (Application) ELISA
Gene ID (Entrez) 5627
Gene Symbol PROS1
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym EC 3.4.21, PROS, protein S (alpha), protein Sa, PS21, PS22, PS23, PS24, PS25, PSA, vitamin K-dependent plasma protein S, vitamin K-dependent protein S
Target Monkey
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.