Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Monkey Triiodothyronine ELISA Kit (Colorimetric)
SDP

Catalog No. NB601761KT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB601761KT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB601761KT Supplier Novus Biologicals™ Supplier No. NBP2601761KIT
Add to Cart
Add to Cart

HAV antibody (IgM) Colorimetric ELISA Kit for study of Infection

The Monkey Triiodothyronine ELISA Kit (Colorimetric) measures monkey Triiodothyronine in serum, plasma, tissue homogenates.

Specifications

Assay Range 0.5 to 8ng/mL
Assay Sensitivity 0.5ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma, Serum, Tissue Homogenates
Specificity Detection of monkey T3
Target Species Monkey
For Use With (Application) ELISA
Kit Contents Assay plate, Standard, Conjugate, HRP-avidin, Wash Buffer (20X concentrate), Substrate A, Substrate B, Stop Solution, Adhesive Strip (For 96 wells), Instruction manual
Quantity 1 Kit
Regulatory Status RUO
Research Discipline Signal Transduction
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Synonym liothyronine; T3
Target Triiodothyronine
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.